Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium nitrate ≥99.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

014493-30 014493.A7 014493-14 014493-A1
AA14493-A7EA 412.41 USD InStock
AA14493-A7 AA14493-30 AA14493-A1 AA14493-14
Sodium nitrate ≥99.0% ACS
Sodium nitrate
In manufacturing of HNO₃, as a catalyst to manufacture H₂SO₄, in manufacturing of glass, enamel for pottery, sodium nitrite


Formula: NaNO₃
MW: 84.99 g/mol
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 308 °C
Density: 2.26
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1,III
REACH: 01-2119488221-41
Merck Index: 13,08720
Order Now
Specification Test Results
Insoluble matter 0.005%
pH of a 5 % solution 5.5-8.3 at 25°
Cl 0.001%
IO3 5 ppm
NO2 0.001%
PO3 5 ppm
SO3 0.003%
Ca 0.005%
Mg 0.002%%
Heavy Metals (as Pb) 5ppm ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty