Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium sulfamate ≥98% ACS

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Ammonium sulfamidate

014534-A3 14534-A3 14534-36 014534-36
AA14534-36EA 114.53 USD InStock
CAAA14534-A3 AA14534-36 AA14534-A3 CAPRAA14534-36
Ammonium sulfamate ≥98% ACS
Ammonium sulphamate
In electroplating solutions, making nitrous oxide gas, for manufacture of flameproofing textiles and paper products


Formula: H₆N₂O₃S
MW: 114.13 g/mol
Boiling Pt: 160 °C (dec.)
Melting Pt: 131…135 °C
Density: >1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011429
CAS Number: 7773-06-0
EINECS: 231-871-7
Merck Index: 12,00589
Order Now
Specification Test Results
Insoluble matter 0.02%
Residue after ignition 0.1%
Heavy Metals (as Pb) 5 ppm
Melting point 133.0° (2deg)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty