Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium chloride, ultra dry ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

14540-30 14540-18 14540-18 14540-09 14540-09 14540-30
AA14540-18EA 255.99 USD InStock
AA14540-18 CAAA14540-09 CAAA14540-18 AA14540-30 CAAA14540-30 AA14540-09
Lithium chloride, ultra dry ≥99.9% (metals basis)
Lithium chloride


Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1325…1360 °C
Melting Pt: 605 °C
Density: 2.068
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty