Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ytterbium (III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

14572-18 014572-18 14572-09 014572-09
AA14572-18EA 248.54 USD InStock
AA14572-18 CAAA14572-09 CAAA14572-18 AA14572-09
Ytterbium (III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Ytterbium (III) sulphate octahydrate


Formula: O₁₂S₃Yb₂·8H₂O
MW: 778.396 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149887
CAS Number: 10034-98-7
EINECS: 236-727-7
Merck Index: 14,10105
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty