Synonyms:
Tantalum(V) ethoxide
14643.18
14643-09
14643-18
14643-03
14643.09
14643.03
AA14643-03EA
62.09
USD
InStock
CAAA14643-03
AA14643-03
AA14643-09
CAAA14643-18
AA14643-18
CAAA14643-09
Tantalum(V) ethanolate ≥99.999% (metals basis) (max. 100 ppm Nb)
Tantalum(V) ethanolate
Decomposes in water. Soluble in organic solvents