Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Heptanesulfonic acid sodium salt monohydrate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium 1-heptanesulphonate monohydrate

15214-18 15214-09 015214-09 015214-18
AA15214-09EA 67.74 USD InStock
AA15214-09 CAAA15214-09 AA15214-18 CAAA15214-18
1-Heptanesulfonic acid sodium salt monohydrate 99%
1-Heptanesulphonic acid sodium salt monohydrate


Formula: C₇H₁₅NaO₃S·1H₂O
MW: 220.27 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149550
CAS Number: 207300-90-1
EINECS: 245-210-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty