Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Europium(III) oxalate hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

15288-09 15288.09 15288.04 15288-04
AA15288-09EA 184.99 USD InStock
AA15288-09 AA15288-04 CAAA15288-04 CAAA15288-09
Europium(III) oxalate hydrate ≥99.9% (REO, rare earth oxide basis)
Europium(III) oxalate hydrate


Formula: C₆Eu₂O₁₂.xH₂O
MW: 567.98 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150143
CAS Number: 152864-32-9
EINECS: 221-885-1
UN: 3288
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty