Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Europium trinitrate hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Europium (III) nitrate hexahydrate

015290-04 15290-04 15290.18 015290-09 15290-18 15290-09
AA15290-09EA 267.99 USD InStock
AA15290-09 AAA15290-18 AA15290-04 CAAA15290-09 CAAAA15290-18 CAAA15290-04
Europium trinitrate hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Europium trinitrate hexahydrate


Formula: EuN₃O₉·6H₂O
MW: 446.07 g/mol
Melting Pt: 85 °C
Density: 2,258 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149700
CAS Number: 10031-53-5
EINECS: 233-380-3
UN: 1477
ADR: 5.1,III
Order Now
Specification Test Results
Form Crystalline

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty