Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cerium triacetate sesquihydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Acetic acid cerium (III) salt sesquihydrate Cerium (III) acetate sesquihydrate

015294-22 015294-36 15294-36 15294-22
AA15294-36EA 424.99 USD InStock
AA15294-36 CAAA15294-36 AA15294-22 CAAA15294-22
Cerium triacetate sesquihydrate ≥99.9% (REO, rare earth oxide basis)
Cerium triacetate sesquihydrate
With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols.

Cerium(III) acetate sesquihydrate, acts as a source of cerium involved in the preparation of cerium oxide. It finds application in metallurgy, glass and glass polishing, ceramics, catalysts, and in phosphors. It is utilized in steel production to remove the free sulfur and oxygen. Further, it is an efficient glass polishing agent.


Formula: C₆H₉CeO₆₄·1,5H₂O
MW: 344.28 g/mol (317.26 anhy)
Melting Pt: 308 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051060
CAS Number: 17829-82-2
EINECS: 208-654-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty