Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methacrylic acid 99+% stabilized

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Methylpropenoic acid

Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) 250 ppm

15546-A1 15546-30 15546-A5 015546-30 015546-A5 015546-A1
AA15546-30EA 40.83 USD InStock
CAAA15546-30 AA15546-30 CAAA15546-A1 AA15546-A1 CAAA15546-A5 AA15546-A5
Methacrylic acid 99+% stabilized
Methacrylic acid


Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 163 °C
Melting Pt: 16 °C
Density: 1.015
Flash Pt: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002651
CAS Number: 79-41-4
EINECS: 201-204-4
UN: 2531
ADR: 8,II
Merck Index: 13,05967
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty