Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Octanesulfonic acid sodium salt monohydrate 99+%

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium 1-octansulphonate monohydrate

015764-09 015764-18 15764-09 15764-18
AA15764-09EA 76.06 USD InStock
AA15764-09 CAAA15764-18 AA15764-18 CAAA15764-09
1-Octanesulfonic acid sodium salt monohydrate 99+%
1-Octanesulphonic acid sodium salt monohydrate


Formula: C₈H₁₇NaO₃S·1H₂O
MW: 234.29 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 207596-29-0
EINECS: 226-195-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty