Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oxalacetic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-succinic acid

15789-09 015789-04 15789-04 015789-09
AA15789-09EA 140.19 USD InStock
CAAA15789-09 AA15789-09 CAAA15789-04 AA15789-04
Oxalacetic acid 98+%
Oxalacetic acid


Formula: C₄H₄O₅
MW: 132.07 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 161 °C (dec.)
Flash Pt: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002592
CAS Number: 328-42-7
EINECS: 206-329-8
UN: 3261
ADR: 8,II
Merck Index: 12,07041
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty