Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Beryllium sulfate tetrahydrate ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

16104-14 016104-22 016104-14 16104-22
AA16104-22EA 447.99 USD InStock
CAAA16104-22 CAAA16104-14 AA16104-22 AA16104-14
Beryllium sulfate tetrahydrate ≥99.99% (metals basis)
Beryllium sulphate tetrahydrate


Formula: BeO₄S·4H₂O
MW: 177.14 g/mol
Boiling Pt: 580 °C (1013 hPa)
Melting Pt: 110 °C
Density: 1.1713
Storage Temperature: Ambient
MDL Number: MFCD00149156
CAS Number: 7787-56-6
EINECS: 236-842-2
UN: 1566
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty