Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-α-Terpineol ≥96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(4-Methylcyclohex-3-en-1-yl)propan-2-olTerpineol

16285.36 16285.22 16285-14 16285-36 16285-22
AA16285-22EA 55.99 USD InStock
AA16285-22 AA16285-36 AA16285-14 CAAA16285-36 CAAA16285-22
(±)-α-Terpineol ≥96%
(±)-α-Terpineol
A naturally-occuring monoterpene alcohol

A naturally-occuring monoterpene alcohol


Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 210…219 °C
Melting Pt: 31…32 °C
Density: 0,930
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001557
CAS Number: 98-55-5
EINECS: 202-680-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty