Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gold(I) iodide 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Gold monoiodide

016617-03 16617-03 16617-06 016617-06
AA16617-03EA 185 USD InStock
CAAA16617-03 AA16617-03 CAAA16617-06 AA16617-06
Gold(I) iodide 99%
Gold (I) iodide


Formula: AuI
MW: 323.87 g/mol
Melting Pt: 120 °C (dec.)
Density: 8,25
Storage Temperature: Ambient
MDL Number: MFCD00014175
CAS Number: 10294-31-2
EINECS: 233-656-3
UN: 3260
ADR: 8,III
Merck Index: 12,04530
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty