Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl ether, anhydrous ≥99% stabilized ACS

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: 2,2'-Oxybisethane2,2'-OxydiethaneEtherEthoxyethaneEthyl ether

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

16767-36 016767-62 016767-18 016767-36 16767-62 16767-18
AA16767-36EA 94.09 USD InStock
AA16767-36 CAAA16767-18 CAAA16767-36 CAAA16767-62 AA16767-62 AA16767-18
Diethyl ether, anhydrous ≥99% stabilized ACS
Diethyl ether


Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 34.6 °C
Melting Pt: –116 °C
Density: 0.713
Flash Pt: –45 °C (–49 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://us-legacy.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg
Order Now
Specification Test Results
Color (APHA) 10
Peroxide (as H2O2) 1ppm ppm
Evaporation residue 0.001%
Titratable acid 0.0002meq/g
Carbonyl (as HCHO) 0.001%
Alcohol (CH3CH2OH) Pass test (limit about 0.05%)
H2O 0.03%

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty