Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Strontium chloride, anhydrous ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

016790-22 016790-14 16790-22 16790-14 16790-36 16790-36
AA16790-22EA 303.08 USD InStock
AA16790-22 CAAA16790-22 AA16790-14 AA16790-36 CAAA16790-14 CAAA16790-36
Strontium chloride, anhydrous ≥99.5% (metals basis)
Strontium chloride


Formula: Cl₂Sr
MW: 158.53 g/mol
Boiling Pt: 1250 °C
Melting Pt: 873 °C
Density: 3.052
Storage Temperature: Ambient
MDL Number: MFCD00011249
CAS Number: 10476-85-4
EINECS: 233-971-6
Merck Index: 12,09000
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty