Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Arsenic (II) sulfide tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

017284-14 017284-06 17284-06 17284-14
AA17284-14EA 172.99 USD InStock
CAAA17284-14 CAAA17284-06 AA17284-14 AA17284-06
Arsenic (II) sulfide tech. 90%, Technical Grade
Arsenic (II) sulphide


Formula: As₂S₂
MW: 213.98 g/mol
Boiling Pt: 565 °C
Melting Pt: ∼307 °C
Density: 3.5
Storage Temperature: Ambient
MDL Number: MFCD00049841
CAS Number: 1303-32-8
UN: 1557
ADR: 6.1,III
Merck Index: 12,00834
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty