Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triallyl phosphite

Supplier: Thermo Scientific Chemicals
Danger

17484-30 17484.09 17484-09 17484-18 17484.18 17484.3
AA17484-09EA 80.99 USD InStock
CAAA17484-30 AA17484-09 CAAA17484-09 AA17484-18 CAAA17484-18 AA17484-30
Triallyl phosphite
Triallyl phosphite


Formula: C₉H₁₅O₃P
Boiling Pt: 84 °C (12 mmHg)
Flash Pt: 86 °C (187 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014948
CAS Number: 102-84-1
EINECS: 203-060-8
UN: 3278
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty