Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper selenide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Copper(II) selenide

17901-14 17901-22 17901-06 17901-22 017901-06 017901-14
AA17901-14EA 348.99 USD InStock
CAAA17901-22 CAAA17901-06 AA17901-14 AA17901-06 AA17901-22 CAAA17901-14
Copper selenide ≥99.5% (metals basis)
Copper selenide
In semiconductors, as catalyst in Kjeldahl digestion


Formula: CuSe
MW: 142.51 g/mol
Melting Pt: 550 °C (dec.)
Density: 6,0…6.6
Storage Temperature: Ambient
MDL Number: MFCD00016058
CAS Number: 1317-41-5
EINECS: 215-272-8
UN: 3077
ADR: 9,III
Merck Index: 12,02719
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty