Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triammonium dodecamolybdophosphate hydrate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Triammonium 12-molybdophosphate hydrate Ammonium phosphomolybdate hydrate

17984-22 017984-22 17984-36 017984-36
AA17984-36EA 277.99 USD InStock
AA17984-36 CAAA17984-36 AA17984-22 CAAA17984-22
Triammonium dodecamolybdophosphate hydrate, REacton®, Reagent Grade
Triammonium dodecamolybdophosphate hydrate


Formula: H₁₂Mo₁₂N₃O₄₀P·H₂O
MW: 1894.36 g/mol
Melting Pt: Dec.
Density: 3.15
Storage Temperature: Ambient
MDL Number: MFCD00036312
CAS Number: 54723-94-3
EINECS: 234-714-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty