Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc citrate dihydrate (≥31% Zn), Reagent Grade

Supplier: Thermo Scientific Chemicals

18129.3 18129-A1 018129.A1 18129-30
AA18129-A1EA 154.99 USD InStock
CAAA18129-30 AA18129-A1 AA18129-30 CAAA18129-A1
Zinc citrate dihydrate (≥31% Zn), Reagent Grade
Zinc citrate dihydrate
Soluble in dilute mineral acids and alkali hydroxides. Slightly soluble in water


Formula: C₁₂H₁₄O₁₆Zn₃
Melting Pt: 334 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150587
CAS Number: 5990-32-9
EINECS: 208-901-2
Merck Index: 14,10134
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty