Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium carbamate 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Carbamic acid ammonium salt

18134.3 18134-18 18134-30 18134.18
AA18134-18EA 50.84 USD InStock
CAAA18134-18 AA18134-18 CAAA18134-30 AA18134-30
Ammonium carbamate 98%
Ammonium carbamate
Ammoniating agent
Freely soluble in water. Soluble in alcohol


Formula: CH₆N₂O₂
MW: 78.07 g/mol
Melting Pt: 60 °C (subl.)
Density: 1.6
Storage Temperature: Ambient
MDL Number: MFCD00013010
CAS Number: 1111-78-0
EINECS: 214-185-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty