Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrafluoroboric acid diethyl ether complex (50 - 55% HBF₄) (w/w)

Supplier: Thermo Scientific Chemicals
Danger

018208.AK 18208-AK 018208.AC 18208-AC
AA18208-ACEA 95.79 USD InStock
AA18208-AC CAAA18208-AC AA18208-AK CAAA18208-AK
Tetrafluoroboric acid diethyl ether complex (50 - 55% HBF₄) (w/w)
Tetrafluoroboric acid-diethyl ether complex (50-55% tetrafluoroboric acid in diethyl ether)
Reacts in water


Formula: C₄H₁₁BF₄O
MW: 161.93 g/mol
Density: 1.19
Flash Pt: 36 °C (97 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00011345
CAS Number: 67969-82-8
EINECS: 267-983-8
UN: 2920
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty