Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,5-Hexadiyne 50% in pentane

Supplier: Thermo Scientific Chemicals
Danger

018720-04 18720-04 018720-09 18720-09
AA18720-09EA 448.99 USD InStock
AA18720-09 CAAA18720-09 CAAA18720-04 AA18720-04
1,5-Hexadiyne 50% in pentane
1,5-Hexadiyne


Formula: C₆H₆
MW: 78.11 g/mol
Density: 0.805
Flash Pt: –2 °C (29 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014924
CAS Number: 628-16-0
EINECS: 211-029-5
UN: 3295
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty