Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetracyclohexyltin

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tetracyclohexylstannane

018822-06 18822-03 18822-06 018822-03 18822-14 018822-14
AA18822-06EA 363.99 USD InStock
AA18822-06 CAAA18822-06 AA18822-03 AA18822-14 CAAA18822-03 CAAA18822-14
Tetracyclohexyltin
Tetracyclohexyltin


Formula: C₂₄H₄₄Sn
MW: 451.32 g/mol
Melting Pt: 241…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00014278
CAS Number: 1449-55-4
EINECS: 215-910-5
UN: 3146
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty