Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron(III) meso-tetraphenylporphine chloride

Supplier: Thermo Scientific Chemicals
Synonyms: 5,10,15,20-Tetraphenyl-21H,23H-porphine Iron(III) chloride5,10,15,20-Tetraphenyl-21H,23H-porphine iron(III) chloridemeso-Tetraphenylporphyrin iron(III) chlorideiron(III) chloride TPPIron(III) Tetraphenylporphyrin chloride21H,23H-Porphine, 5,10,15,20-tetraphenyl-, iron complex

018829.MD 18829-MD 18829-03 18829.03
AA18829-MDEA 56.19 USD InStock
CAAA18829-03 AA18829-MD CAAA18829-MD AA18829-03
Iron(III) meso-tetraphenylporphine chloride
Iron(III) meso-tetraphenylporphine chloride


Formula: C₄₄H₃₀Cl₃FeN₄
MW: 704.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012152
CAS Number: 16456-81-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty