Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyridine ≥99.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

19378-AP 019378-K7 19378-M1 019378-M1 019378-K2 019378-AP 19378-K2 19378-K7
AA19378-K2EA 265.41 USD InStock
CAAA19378-K7 AA19378-K2 CAAA19378-AP AA19378-K7 CAAA19378-M1 AA19378-AP CAAA19378-K2 AA19378-M1
Pyridine ≥99.0% ACS
Pyridine


Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115 °C
Melting Pt: –42 °C
Density: 0.978
Flash Pt: 20 °C (68 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II
Merck Index: 13,08060
https://us-legacy.vwr.com/stibo/web/std.lang.all/07/97/790797.jpg
Order Now
Specification Test Results
Evaporation residue 0.002%
H2O 0.1%
Cl 0.001%
SO 0.00001
NH3 0.002%
Cu 5 ppm
Reducing substances Pass test
Solubility in water Pass test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty