Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) acetate, anhydrous

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cupric acetate Copper diacetate Acetic acid copper (II) salt

19417-14 19417-36 19417-22 019417-22 19417.36 019417-14
AA19417-22EA 178.99 USD InStock
CAAAA19417-36 AA19417-22 CAAA19417-22 AAA19417-36 AA19417-14 CAAA19417-14
Copper(II) acetate, anhydrous
Copper (II) acetate
Anhydrous


Formula: CuCH₃CO₂
MW: 181.64 g/mol
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008690
CAS Number: 142-71-2
EINECS: 205-553-3
UN: 3077
ADR: 9,III
Merck Index: 12,02690
Order Now
Specification Test Results
Form Powder

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty