Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Cyclohexene-1-acetic acid

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Cyclohexenylacetic acid

19462-06 19462-14 19462-03 19462.06 019462-03 19462.14
AA19462-06EA 113.99 USD InStock
AA19462-06 CAAA19462-06 AA19462-03 AA19462-14 CAAA19462-14 CAAA19462-03
1-Cyclohexene-1-acetic acid
1-Cyclohexene-1-acetic acid


Formula: C₈H₁₂O₂
MW: 140.182 g/mol
Boiling Pt: 140…142 °C (25 mmHg)
Melting Pt: 33 °C
Density: 1,04
Storage Temperature: Ambient
MDL Number: MFCD00015482
CAS Number: 18294-87-6
EINECS: 242-172-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty