Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chromotropic acid disodium salt dihydrate ACS

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4,5-Dihydroxy-2,7-naphthalenedisulphonic acid disodium salt dihydrate 4,5-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt dihydrate Disodium 4,5-dihydroxynaphthalene-2,7-disulphonate dihydrate

022414-14 22414-14 022414-06 022414-22 22414-06
AA22414-06EA 49.42 USD InStock
AA22414-06 CAAA22414-06 AA22414-14 CAAA22414-14 AA22414-22
Chromotropic acid disodium salt dihydrate ACS
Chromotropic acid disodium salt dihydrate


Formula: (HO)₂C₁₀H₄(SO₃Na)₂·2H₂O
MW: 400.29 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150612
CAS Number: 5808-22-0
EINECS: 204-972-9
Merck Index: 13,02261
Order Now
Specification Test Results
Solution Pass Test
Appearance Off-white to buff-colored
Sensitivity to nitrate Pass Test
Sensitivity to formaldehyde Pass test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty