Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium metaphosphate ≥90%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Bariummetaphosphate

22641-36 22641.36
AA22641-36EA 215.99 USD InStock
CAAA22641-36 AA22641-36
Barium metaphosphate ≥90%
Barium metaphosphate
Typically 95% phosphate assay

Insoluble in water. Soluble in acids by a slow dissolution


Formula: Ba(PO₃)₂
MW: 295.25 g/mol
Melting Pt: 1560 °C
Storage Temperature: Ambient
MDL Number: MFCD00061417
CAS Number: 13762-83-9
EINECS: 237-362-6
UN: 1564
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty