Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tricadmium diarsenide ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cadmium arsenide

22773-09 022773-09
AA22773-09EA 189.99 USD InStock
CAAA22773-09 AA22773-09
Tricadmium diarsenide ≥99% (metals basis)
Tricadmium diarsenide


Formula: As₂Cd₃
MW: 487.07 g/mol
Melting Pt: 715 °C
Density: 3,031
Storage Temperature: Ambient
MDL Number: MFCD00049723
CAS Number: 12006-15-4
EINECS: 234-484-1
UN: 1557
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty