Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylacetamide ≥99.5% for HPLC

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Acetic acid dimethylamide

022916.M1 022916.K7 022916.K2
AA22916-K7EA 432.19 USD InStock
AA22916-K7 AA22916-M1 AA22916-K2
N,N-Dimethylacetamide ≥99.5% for HPLC
N,N-Dimethylacetamide


Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 165…166 °C
Melting Pt: –20 °C
Density: 0.940
Flash Pt: 70 °C (158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
UN: 2810
ADR: 6.1,III
Merck Index: 13,03254
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/13/2298913.jpg
Order Now
Specification Test Results
Evaporation residue 5 ppm
H2O 0.0005

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty