Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclohexane ≥99% for HPLC

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Hexahydro-BenzeneHexamethylene

22919-K7 22919-M1 022919-K2 022919-M1 22919-K2 022919-K7
AA22919-K7EA 198.12 USD InStock
AA22919-K7 CAAA22919-M1 CAAA22919-K2 AA22919-M1 AA22919-K2 CAAA22919-K7
Cyclohexane ≥99% for HPLC
Cyclohexane


Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 80…81 °C
Melting Pt: 6.5 °C
Density: 0.779
Flash Pt: –18 °C (–1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://us-legacy.vwr.com/stibo/web/std.lang.all/23/06/772306.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty