Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorobenzene ≥99.9% for spectrophotometry

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Phenylchloride

022921.M1 022921.K2 022921.AP 022921.K7
AA22921-APEA 61 USD InStock
AA22921-AP AA22921-K7 AA22921-K2 AA22921-M1
Chlorobenzene ≥99.9% for spectrophotometry
Chlorobenzene


Formula: C₆H₅Cl
MW: 112.56 g/mol
Boiling Pt: 132 °C
Melting Pt: –45 °C
Density: 1.106
Flash Pt: 28 °C (82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://us-legacy.vwr.com/stibo/web/std.lang.all/08/56/750856.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty