Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Molybdic acid ≥85% (MoO₃ basis) ACS

Supplier: Thermo Scientific Chemicals
Warning

22983-A3 22983-36 022983-36 022983-A3
AA22983-A3EA 607.99 USD InStock
CAAA22983-A3 AA22983-A3 CAAA22983-36 AA22983-36
Molybdic acid ≥85% (MoO₃ basis) ACS
Molybdic acid


Formula: H₂MoO₄
MW: 161.95 g/mol
Boiling Pt: ∼300 °C (1013 hPa)
Melting Pt: 300 °C
Density: 3.1
Storage Temperature: Ambient
MDL Number: MFCD00036287
CAS Number: 7782-91-4
EINECS: 231-970-5
Merck Index: 12,06322
Order Now
Specification Test Results
Insoluble in dilute ammonium hydroxide 0.01%
Cl 0.002%
Arsenate phosphate and silicate (as SiO2) 0.001%
PO3 5 ppm
SO3 0.2%
Heavy Metals (as Pb) 0.003%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty