Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chromium(III) phosphate tetrahydrate

Supplier: Thermo Scientific Chemicals
Warning

24110.09 24110-18 24110.18 24110-09
AA24110-09EA 43.99 USD InStock
AA24110-09 CAAA24110-09 AA24110-18 CAAA24110-18
Chromium(III) phosphate tetrahydrate
Chromium(III) phosphate tetrahydrate
Applications: Catalyst, pigment


Formula: CrH₈O₈P
Density: 2,26
Storage Temperature: Ambient
MDL Number: MFCD00016049
CAS Number: 10101-59-4
EINECS: 232-141-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty