Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloro(methyl)phosphine 97+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Methyldichlorophosphine

030211-06
AA30211-06EA 357.99 USD InStock
AA30211-06
Dichloro(methyl)phosphine 97+%
Dichloro(methyl)phosphine


Formula: CH₃Cl₂P
MW: 116.91 g/mol
Boiling Pt: 81…82 °C (729 mmHg)
Density: 1.309
Flash Pt: 48 °C (120 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013616
CAS Number: 676-83-5
EINECS: 211-631-8
UN: 2845
ADR: 4.2,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty