Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

030749-A5 030749-30 030749-A1
AA30749-A1EA 52 USD InStock
AA30749-A1 AA30749-30 AA30749-A5
Benzaldehyde 98%
Benzaldehyde


Formula: C₆H₅CHO
MW: 106.12 g/mol
Boiling Pt: 178…179 °C
Melting Pt: –26 °C
Density: 1.047
Flash Pt: 62 °C (143 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003299
CAS Number: 100-52-7
EINECS: 202-860-4
UN: 1990
ADR: 9,III
Merck Index: 13,01057
https://us-legacy.vwr.com/stibo/web/std.lang.all/60/95/746095.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty