Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetramethylammonium hydroxide 25% (w/w) in methanol

Supplier: Thermo Scientific Chemicals
Danger

030833-09 30833-36 030833-22 30833-22 030833-36 30833-09
AA30833-22EA 69.35 USD InStock
AA30833-22 CAAA30833-36 CAAA30833-22 AA30833-36 AA30833-09 CAAA30833-09
Tetramethylammonium hydroxide 25% (w/w) in methanol
Tetramethylammonium hydroxide


Formula: C₄H₁₃NO
MW: 91.15 g/mol
Density: 0.865
Flash Pt: 12 °C (54 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 2924
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty