Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron selenide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

031112-09 31112-09 31112-18 031112-18
AA31112-09EA 180.99 USD InStock
CAAA31112-18 AA31112-09 CAAA31112-09 AA31112-18
Iron selenide ≥99.9% (metals basis)
Iron selenide


Formula: FeSe
MW: 134.81 g/mol
Melting Pt: 965 °C
Density: 4,72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016085
CAS Number: 1310-32-3
EINECS: 215-177-1
UN: 3283
ADR: 6.1,III
Merck Index: 12,04103
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty