Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-1,10-phenanthroline

Supplier: Thermo Scientific Chemicals

031180-04 031180-02 31180-04 31180-02
AA31180-02EA 81.79 USD InStock
AA31180-02 CAAA31180-02 AA31180-04 CAAA31180-04
5-Chloro-1,10-phenanthroline
5-Chloro-1,10-phenanthroline


Formula: C₁₂H₇ClN₂
Melting Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD00004980
CAS Number: 4199-89-7
EINECS: 224-098-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty