Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol absolute ≥99.8% ACS low acetone

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: CarbinolMethyl alcoholMethyl hydroxideMethylolWood alcohol

31721-K2 031721-M1 031721-K7 031721-K2 031721-AP 31721-K7 31721-AP 31721-M1
AA31721-APEA 29.3 USD InStock
AA31721-AP CAAA31721-M1 AA31721-K7 CAAA31721-AP AA31721-K2 CAAA31721-K7 AA31721-M1 CAAA31721-K2
Methanol absolute ≥99.8% ACS low acetone
Methanol


Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.7 °C
Melting Pt: –98 °C
Density: 0.791
Flash Pt: 11 °C (52 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
Appearance Clear
Color (APHA) 10
H2O 0.1%
Evaporation residue 0.001%
Solubility in water Pass test
Carbonyl compounds 0.001 % each of acetone formaldehyde and acetaldehyde
Titratable acid 0.0003meq/g
Titratable base 0.0002meq/g

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty