Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Hexanesulfonic acid sodium salt monohydrate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium 1-hexanesulphonate monohydrate

032190-09 32190-18 32190-09 032190-18
AA32190-18EA 343.92 USD InStock
CAAA32190-18 CAAA32190-09 AA32190-18 AA32190-09
1-Hexanesulfonic acid sodium salt monohydrate 99%
1-Hexanesulphonic acid sodium salt monohydrate


Formula: C₆H₁₃NaO₃S·1H₂O
MW: 206.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149549
CAS Number: 207300-91-2
EINECS: 220-601-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty