Would you like to log in again?
Ceramic Multibore Insulating Tubes, AL23, Round, Thermo Scientific
Supplier: Thermo Scientific
32552-FB 32552-FB 32558-FB 32554-FB 32554-EG 32558-DZ 32550-DS 32558-EW 32560-ER 32552-EW 32556-FB 32550-FB 32558-ER 32558-DS 32560-DZ 32552-EL 32554-DZ 32556-EW 32552-EW 32552-ER 32550-ER 32550-FB 32556-DV 32560-EW 32550-DZ 32556-DS 32552-DV 32560-EW 32554-DS 032554.EW 32556-DS 32554-EW 32558-DZ 32550-EW 32558-FB 32550-EL 32550-DV 32552-EG 32550-DV 32552-DZ 32554-EL 32560-DV 32550-ER 32554-DS 32556-DZ 32556-FB 32550-EG 32556-EL 32558-EL 32552-DS 32550-EL 32560-DS 32552-ER 32552-DS 32560-FB 32560-EL 32560-DS 32556-EW 32560-FB 32554-ER 32556-EG 32558-DV 32560-EL 32560-EG 32550-EW 032560.DZ 32554-EG 32552-DV 32552-DZ 32556-DZ 32560-DV 32554-EL 32556-EG 32554-DV 32560-ER 32558-DV 32558-DS 32558-ER 32560-EG 32558-EW 032556.ER 032556.DV 32550-DS 32558-EG 32556-ER 32554-DV 32550-EG 32554-DZ 32554-FB 32550-DZ 32552-EG 32552-EL 032558.EL 32558-EG 32556-EL 32554-ER
AA32560-FBEA 346.34 USD InStock
AA32560-FB AA32550-ER CAAA32552-EL CAAA32556-EL AA32550-EW CAAA32552-EG AA32554-EW AA32550-EL CAAA32556-EG AA32554-ER AA32558-EW AA32558-ER CAAA32552-DZ AA32550-FB AA32560-ER CAAA32556-DZ CAAA32552-DV AA32560-EW CAAA32556-DV AA32554-FB CAAA32552-DS CAAA32556-DS AA32560-EL AA32558-FB AA32552-DV AA32556-DZ AA32552-DS AA32556-DV CAAA32552-FB AA32556-DS CAAA32556-FB AA32552-EG AA32556-EL CAAA32552-EW AA32556-EG CAAA32556-EW CAAA32552-ER AA32552-DZ CAAA32556-ER AA32552-EW CAAA32554-EL CAAA32560-ER CAAA32558-ER CAAA32560-EW CAAA32554-EG AA32556-EW CAAA32558-EL CAAA32550-EG CAAA32558-EG AA32552-ER CAAA32560-EL CAAA32550-DZ AA32552-EL AA32556-ER CAAA32554-DZ CAAA32550-DS CAAA32554-DV CAAA32558-DZ CAAA32550-DV CAAA32554-DS AA32552-FB CAAA32558-DV AA32556-FB CAAA32560-FB CAAA32558-DS AA32550-DS CAAA32560-DS AA32554-DZ CAAA32560-DV AA32558-DZ AA32550-DV AA32560-EG AA32554-DV AA32558-DV AA32554-DS AA32560-DZ AA32558-DS CAAA32550-FB CAAA32554-FB CAAA32558-FB AA32554-EL CAAA32560-EG AA32558-EL CAAA32554-EW AA32550-EG AA32560-DS AA32560-DV CAAA32550-EW AA32554-EG CAAA32560-DZ AA32550-DZ AA32558-EG CAAA32550-EL CAAA32558-EW CAAA32550-ER CAAA32554-ER
Ceramic Multibore Insulating Tubes, AL23, Round, Thermo Scientific
Tubing Transfer Tubing
Pure Al₂O₃ Aluminium oxide, AL23.
  • Corrosion resistant
  • Permeable to microwaves
Dense, excellent thermal and electrical resistance properties.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty