Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) hydroxide 94% stabilized, tech.

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Copper dihydroxide Cupric hydroxide

Stabilization: Stabilised with copper

32733-A3 32733-36 032733-A3 032733-36
AA32733-36EA 92.12 USD InStock
AA32733-36 CAAA32733-36 AA32733-A3 CAAA32733-A3
Copper(II) hydroxide 94% stabilized, tech.
Copper (II) hydroxide
Used as a raw material in chemical processing and catalyst manufacturing.


Formula: H₂CuO₂
MW: 97.56 g/mol
Density: 3.4
Storage Temperature: Ambient
MDL Number: MFCD00010968
CAS Number: 20427-59-2
EINECS: 243-815-9
UN: 3288
ADR: 6.1,II
Merck Index: 12,02709
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty