Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-O-Palmitoyl-L-ascorbic acid 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Ascorbyl PalmitatePalmitoyl-L-ascorbic acid

032791-14 32791-36 32791-14 032791-36 32791-22 032791-22
AA32791-22EA 136.05 USD InStock
CAAA32791-36 CAAA32791-14 AA32791-22 CAAA32791-22 AA32791-14 AA32791-36
6-O-Palmitoyl-L-ascorbic acid 99%
6-O-Palmitoyl-L-ascorbic acid


Formula: C₂₂H₃₈O₇
MW: 414.54 g/mol
Boiling Pt: ∼250 °C
Melting Pt: 113…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00005377
CAS Number: 137-66-6
EINECS: 205-305-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty