Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ceramic Rods, Round, Solid, AL23, Thermo Scientific
Supplier: Thermo Scientific
32937-EO 32923-DT 032929-EO 32937-EH 032933-DW 032931-DR 032925-DR 32937-EH 32920-DT 32933-EH 32923-DR 32925-DT 032926-DT 32927-EM 32933-EM 032927-EH 032922-DT 32935-QAT 032922-DR 32933-EO 32933-DT 32929-DR 032922-DW 032929-DM 32937-DW 32935-DR 32931-EM 032925-DM 32920-DR 032924-DW 32929-DW 32929-EM 032933-EM 032931-DW 32926-QAT 32935-DW 32926-DW 32920-DW 32925-DW 032923-DW 32937-DW 032931-QAT 32924-DR 32925-DW 032927-EO 32924-DM 32927-DW 32927-DR 32923-DR 32927-EM 32927-DT 32927-DW 32935-DW 32924-DM 32927-EO 32922-DM 32922-DT 32926-DM 032921-DM 032927-QAT 32929-DT 32929-EO 32937-DT 32933-DM 32935-DT 32937-DM 32931-EM 32921-DR 32931-EO 32929-EH 32921-DR 32920-DT 32931-DW 32933-DR 32925-DR 32935-EH 32933-QAT 32922-DR 032937-EM 32924-DT 32923-DT 032926-DM 32920-DM 32931-DR 32924-DR 32929-EH 32922-DW 32933-EH 032933-DR 32926-DT 032920-DM 32927-DR 32929-DW 32926-QAT 32931-QAT 32927-QAT 32929-DR 32925-QAT 32935-EM 32937-DM 32925-QAT 32921-DM 32933-QAT 32937-DR 32935-EO 32923-DW 032929-QAT 32921-DW 32937-EM 32927-DM 32931-EH 32935-DT 32925-DT 32935-EH 032931-EH 032931-DT 32929-EM 032931-DM 32935-EO 32935-DM 32933-DW 32921-DW 32926-DR 32923-DM 32926-DR 32929-QAT 32927-DT 32937-QAT 32931-DM 32927-EH 032920-DR 32937-DR 032922-DM 32935-EM 032929-DT 32933-DT 32931-DT 32937-QAT 32931-EO 32923-DM 32937-EO 32929-DM 32926-DW 32937-DT 32924-DW 32935-QAT 32924-DT 32925-DM 32927-DM 32933-EO 032920-DW 032933-DM 32935-DM
AA32931-DREA 45.09 USD InStock
AA32931-DR CAAA32924-DW AA32931-DT AA32925-DR AA32931-QAT CAAA32933-DM AA32931-DW CAAA32933-DR AA32925-DM CAAA32927-DM AA32922-DW AA32922-DR AA32922-DT CAAA32927-DW CAAA32927-DT AA32931-EH AA32931-EO CAAA32927-DR AA32931-EM AA32927-EM AA32927-EH CAAA32933-EH CAAA32933-EO CAAA32927-EH AA32922-DM CAAA32933-EM AA32927-EO CAAA32927-EO CAAA32933-DT CAAA32929-QAT CAAA32927-EM AA32927-DW CAAA32933-DW AA32937-DT CAAA32922-DR AA32937-DR AA32925-QAT AA32937-DM CAAA32922-DM AA32927-DM CAAA32922-DW AA32937-DW CAAA32922-DT AA32927-DT AA32927-DR CAAA32931-DM AA32937-EM CAAA32935-QAT CAAA32925-DM AA32924-DW AA32937-EH AA32924-DT CAAA32925-DT CAAA32925-DR AA32937-EO CAAA32931-EH CAAA32925-DW CAAA32931-EM AA32924-DR CAAA32926-QAT AA32929-EM AA32924-DM AA32929-EO CAAA32931-EO CAAA32931-DT AA32921-DW CAAA32931-DR CAAA32931-DW AA32921-DR AA32929-EH CAAA32937-QAT CAAA32920-DM AA32935-DM AA32935-DW CAAA32920-DW AA32929-DR CAAA32937-DM AA32935-DT AA32929-DT CAAA32920-DR AA32929-DW CAAA32920-DT AA32921-DM CAAA32937-DT AA32929-DM CAAA32937-DR AA32935-EH AA32926-DW CAAA32923-DR CAAA32923-DM AA32935-EM CAAA32931-QAT AA32935-EO CAAA32937-EM CAAA32937-EO CAAA32923-DT CAAA32937-EH CAAA32923-DW AA32926-DT AA32926-DR AA32926-QAT AA32926-DM CAAA32926-DM AA32923-DW AA32923-DT CAAA32937-DW AA32923-DR AA32933-DM CAAA32926-DT CAAA32933-QAT CAAA32926-DR AA32933-DW AA32933-DR AA32933-DT CAAA32926-DW CAAA32935-DR CAAA32935-DT CAAA32935-DM AA32923-DM AA32937-QAT CAAA32929-DR AA32927-QAT AA32920-DT AA32933-EH CAAA32929-DT CAAA32927-QAT CAAA32929-DW AA32920-DR AA32933-EO CAAA32929-DM AA32920-DW AA32933-EM CAAA32921-DM CAAA32921-DW CAAA32935-EM CAAA32921-DR CAAA32929-EH CAAA32935-EH AA32920-DM AA32935-QAT CAAA32935-EO AA32933-QAT AA32925-DW CAAA32935-DW AA32925-DT CAAA32925-QAT CAAA32924-DT AA32929-QAT CAAA32929-EM CAAA32924-DR CAAA32929-EO AA32931-DM CAAA32924-DM
Ceramic Rods, Round, Solid, AL23, Thermo Scientific
Solid Rods
Pure Al2O3 Aluminium oxide, AL23
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Solid aluminum rod.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty