Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cadmium(II) bromide tetrahydrate, Reagent Grade

Supplier: Thermo Scientific Chemicals
Warning

32958-14 032958-14 32958-22 032958-22
AA32958-14EA 45.44 USD InStock
AA32958-14 AA32958-22 CAAA32958-14 CAAA32958-22
Cadmium(II) bromide tetrahydrate, Reagent Grade
Cadmium (II) bromide tetrahydrate


Formula: Br₂Cd·4H₂O
MW: 344.28 g/mol
Melting Pt: 575 °C
Density: 5.214 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150204
CAS Number: 13464-92-1
EINECS: 232-165-1
UN: 2570
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty